Assay Details
| Gene Symbol: Nbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031609.1 | NP_113797.1 | 1 - 2 | 166 | 98 | |
| GenBank mRNA | BC061833.1 | - | 1 - 2 | 113 | 5599381 | 98 |
| S72637.1 | - | 1 - 2 | 166 | 98 | ||
| X66872.1 | - | 1 - 2 | 166 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061833.1, NM_031609.1, S72637.1, X66872.1 transcripts.
Set Membership: |
|

Nbl1
Genomic Map