Assay Details
| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053327.1 | NP_445779.1 | 1 - 2 | 33 | 56 | |
| GenBank mRNA | D13927.1 | - | 1 - 2 | 33 | 56 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the D13927.1,NM_053327.1 transcripts.
Set Membership: |
|

Clcnka
Genomic Map