Assay Details
| Gene Symbol: Cyp21a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057101.2 | NP_476442.2 | 9 - 10 | 1211 | 55 | |
| XM_006255907.2 | XP_006255969.1 | 9 - 10 | 1285 | 55 | ||
| GenBank mRNA | U56853.1 | - | 9 - 10 | 1212 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp21a1
Genomic Map