Assay Details
| Gene Symbol: Abcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133401.1 | NP_596892.1 | 15 - 16 | 1950 | 178 | |
| XM_006235994.2 | XP_006236056.1 | 17 - 18 | 2775 | 178 | ||
| GenBank mRNA | AF257746.1 | - | 15 - 16 | 1950 | 178 | |
| AY582535.1 | - | 14 - 15 | 1865 | 178 | ||
| S66618.1 | - | 2 - 3 | 315 | 178 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb1a
Genomic Map