Assay Details
| Gene Symbol: Akr7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134407.1 | NP_599234.1 | 4 - 5 | 682 | 85 | |
| GenBank mRNA | AB037424.1 | - | 4 - 5 | 682 | 85 | |
| AF503514.1 | - | 4 - 5 | 720 | 85 | ||
| AJ271883.1 | - | 4 - 5 | 685 | 85 | ||
| BC061816.1 | - | 4 - 5 | 685 | 5598724 | 85 | |
| FQ230711.1 | - | 1 - 2 | 91 | 85 | ||
| FQ231883.1 | - | 1 - 2 | 91 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akr7a2
Genomic Map