Assay Details
| Gene Symbol: Nrep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178096.3 | NP_835197.1 | 1 - 2 | 56 | 95 | |
| XM_006254595.1 | XP_006254657.1 | 2 - 3 | 155 | 95 | ||
| GenBank mRNA | AB072344.1 | - | 1 - 2 | 61 | 95 | |
| AY724475.1 | - | 2 - 3 | 87 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB072344.1, NM_178096.3, XM_006254595.1 transcripts.
Set Membership: |
|

Nrep
Genomic Map