Assay Details
| Gene Symbol: Fpgt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199494.1 | NP_955788.1 | 1 - 2 | 79 | 95 | |
| XM_006233519.2 | XP_006233581.1 | 1 - 2 | 105 | 95 | ||
| GenBank mRNA | AJ276066.1 | - | 1 - 2 | 114 | 95 | |
| BC088089.1 | - | 1 - 2 | 81 | 95 | ||
| BC168244.1 | - | 1 - 2 | 62 | 5600080 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fpgt
Genomic Map