Assay Details
| Gene Symbol: Tas2r105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023999.1 | NP_076489.1 | 1 | 547 | 137 | |
| GenBank mRNA | AF227146.1 | - | 1 | 547 | 137 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tas2r105
Genomic Map