Assay Details
| Gene Symbol: Gipc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053341.1 | NP_445793.1 | 3 - 4 | 704 | 93 | |
| XM_006255311.2 | XP_006255373.1 | 3 - 4 | 2969 | 93 | ||
| XM_006255312.2 | XP_006255374.1 | 4 - 5 | 1963 | 93 | ||
| XM_008772420.1 | XP_008770642.1 | 4 - 5 | 1893 | 93 | ||
| GenBank mRNA | AF032120.1 | - | 3 - 4 | 704 | 93 | |
| AF089817.1 | - | 4 - 5 | 722 | 93 | ||
| BC061964.1 | - | 2 - 3 | 1512 | 93 | ||
| BC070505.1 | - | 4 - 5 | 747 | 7097697 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gipc1
Genomic Map