Assay Details
| Gene Symbol: Fgf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130751.1 | NP_570107.1 | 1 - 2 | 189 | 116 | |
| GenBank mRNA | AB078902.1 | - | 1 - 2 | 189 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgf22
Genomic Map