Assay Details
Gene Symbol: Nacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134413.2 | NP_599240.1 | 3 - 4 | 1212 | 68 | |
XM_006255214.2 | XP_006255276.1 | 3 - 4 | 1206 | 68 | ||
XM_006255215.2 | XP_006255277.1 | 3 - 4 | 1126 | 68 | ||
XM_008772346.1 | XP_008770568.1 | 3 - 4 | 1229 | 68 | ||
GenBank mRNA | AF015911.1 | - | 4 - 5 | 1213 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
