Assay Details
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138905.2 | NP_620260.2 | 5 - 6 | 1227 | 133 | |
GenBank mRNA | BC072544.1 | - | 5 - 6 | 1227 | 7105872 | 133 |
Y07783.1 | - | 5 - 6 | 1219 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
gastrulation with mouth forming second
negative regulation of protein phosphorylation
phospholipid metabolic process
cell adhesion
dephosphorylation
single organismal cell-cell adhesion
regulation of Wnt signaling pathway
homotypic cell-cell adhesion
canonical Wnt signaling pathway involved in positive regulation of endothelial cell migration
canonical Wnt signaling pathway involved in positive regulation of cell-cell adhesion
canonical Wnt signaling pathway involved in positive regulation of wound healing
positive regulation of peptidyl-tyrosine phosphorylation
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
Bergmann glial cell differentiation
canonical Wnt signaling pathway
regulation of sphingolipid mediated signaling pathway
integrin binding
phosphatidate phosphatase activity
sphingosine-1-phosphate phosphatase activity
lipid phosphatase activity
blood vessel development
gastrulation with mouth forming second
negative regulation of protein phosphorylation
phospholipid metabolic process
cell adhesion
dephosphorylation
single organismal cell-cell adhesion
regulation of Wnt signaling pathway
homotypic cell-cell adhesion
canonical Wnt signaling pathway involved in positive regulation of endothelial cell migration
canonical Wnt signaling pathway involved in positive regulation of cell-cell adhesion
canonical Wnt signaling pathway involved in positive regulation of wound healing
positive regulation of peptidyl-tyrosine phosphorylation
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
Bergmann glial cell differentiation
canonical Wnt signaling pathway
regulation of sphingolipid mediated signaling pathway
integrin binding
phosphatidate phosphatase activity
sphingosine-1-phosphate phosphatase activity
lipid phosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1007105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98532 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1007105 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1007105 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1007105 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1007105 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13332 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700694946 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31049 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94456 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2307 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77484 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32761 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700694946 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10870485 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870483 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870487 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870484 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870486 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870482 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10870481 | - | - |
Rat Genome 230 2.0 Array | 1370950_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370951_at | - | - |
Rat Genome 230 2.0 Array | 1370951_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370950_at | - | - |
Rat Genome 230 2.0 Array | 1372101_at | - | - |
Rat Genome 230 2.0 Array | 1372101_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370951_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372101_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372101_at | - | - |
Affymetrix HT Rat Focus Array | 1370950_at | - | - |
Affymetrix HT Rat Focus Array | 1370951_at | - | - |
Affymetrix HT Rat Focus Array | 1370950_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500007248.rn.1 | TC0500001110.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500007250.rn.1 | TC0500001110.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500007247.rn.1 | TC0500001110.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500007249.rn.1 | TC0500001110.rn.1 | RTA TC |
Rat Expression Set 230 | 1370950_at | - | - |
Rat Expression Set 230 | 1372101_at | - | - |
Rat Expression Set 230 | 1370951_at | - | - |
Rat Expression Set 230 | 1370951_PM_at | - | - |
Rat Expression Set 230 | 1370950_PM_at | - | - |
Rat Expression Set 230 | 1372101_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870481 | - | - |
Rat Genome U34 Set | rc_AI013472_at | - | - |
Rat Genome U34 Set | rc_AA924925_at | - | - |
Rat Genome U34 Set | rc_AI177031_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21282613 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361266 | - | - |
RatRef-12 Expression BeadChip | ILMN_65901 | - | - |
