Assay Details
Gene Symbol: Cacna1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195199.1 | NP_001182128.1 | 10 - 11 | 1340 | 53 | |
NM_147141.1 | NP_671482.1 | 10 - 11 | 1428 | 53 | ||
XM_006233569.2 | XP_006233631.1 | 10 - 11 | 1471 | 53 | ||
XM_006233570.2 | XP_006233632.1 | 10 - 11 | 1471 | 53 | ||
XM_006233572.2 | XP_006233634.1 | 10 - 11 | 1471 | 53 | ||
XM_006233573.2 | XP_006233635.1 | 10 - 11 | 1471 | 53 | ||
XM_006233574.2 | XP_006233636.1 | 10 - 11 | 1471 | 53 | ||
XM_006233575.2 | XP_006233637.1 | 10 - 11 | 1471 | 53 | ||
XM_006233576.2 | XP_006233638.1 | 10 - 11 | 1471 | 53 | ||
XM_006233577.2 | XP_006233639.1 | 10 - 11 | 1471 | 53 | ||
XM_006233578.2 | XP_006233640.1 | 10 - 11 | 1468 | 53 | ||
XM_008761578.1 | XP_008759800.1 | 10 - 11 | 1471 | 53 | ||
GenBank mRNA | AF055477.2 | - | 10 - 11 | 1428 | 53 | |
HQ008360.1 | - | 10 - 11 | 1340 | 53 | ||
M92905.1 | - | 10 - 11 | 1340 | 53 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
