Assay Details
| Gene Symbol: Bzw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198789.2 | NP_942084.1 | 3 - 4 | 340 | 87 | |
| XM_006245000.2 | XP_006245062.2 | 3 - 4 | 339 | 87 | ||
| GenBank mRNA | BC061580.1 | - | 3 - 4 | 344 | 6890414 | 87 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bzw1
Genomic Map