Assay Details
Gene Symbol: Pgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201990.1 | NP_973719.1 | 26 - 27 | 2643 | 66 | |
GenBank mRNA | AB116149.1 | - | 26 - 27 | 2643 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
intracellular protein transport
sensory perception of sound
embryonic pattern specification
anterior/posterior axis specification
myo-inositol transport
forebrain regionalization
head development
nucleic acid phosphodiester bond hydrolysis
nuclease activity
phosphoric ester hydrolase activity
GPI anchor biosynthetic process
intracellular protein transport
sensory perception of sound
embryonic pattern specification
anterior/posterior axis specification
myo-inositol transport
forebrain regionalization
head development
nucleic acid phosphodiester bond hydrolysis
nuclease activity
phosphoric ester hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P407204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178077 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P407204 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P407204 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P407204 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P407204 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21134 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80428 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36002 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88668 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89378 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10928070 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928071 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928075 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928060 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928078 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928081 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928080 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928067 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928057 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928082 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928063 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928068 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928077 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928073 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928074 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928059 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928076 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928065 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928079 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928058 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928083 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928061 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928066 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928069 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928062 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928072 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928064 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10928056 | - | - |
Affymetrix Rat Clariom S Assay | PSR0900016282.rn.1 | TC0900001987.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900016281.rn.1 | TC0900001987.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10928056 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20961420 | - | - |
ABI Rat Genome Survey Microarray | 22318134 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367321 | - | - |
RatRef-12 Expression BeadChip | ILMN_63959 | - | - |
