Assay Details
| Gene Symbol: Cebpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287577.1 | NP_001274506.1 | 1 | 877 | 149 | |
| NM_001287578.1 | NP_001274507.1 | 1 | 877 | 149 | ||
| NM_001287579.1 | NP_001274508.1 | 1 | 877 | 149 | ||
| NM_012524.3 | NP_036656.1 | 1 | 877 | 149 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cebpa
Genomic Map