Assay Details
Gene Symbol: Ren | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012642.4 | NP_036774.4 | 5 - 6 | 708 | 108 | |
GenBank mRNA | BC078878.1 | - | 5 - 6 | 708 | 7126969 | 108 |
J02941.1 | - | 5 - 6 | 901 | 108 | ||
S60054.1 | - | 5 - 6 | 718 | 108 | ||
X07033.1 | - | 5 - 6 | 718 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
