Assay Details
| Gene Symbol: Rpl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022515.1 | NP_071960.1 | 2 - 3 | 104 | 139 | |
| GenBank mRNA | BC058473.1 | - | 2 - 3 | 129 | 6918141 | 139 |
| FQ209984.1 | - | 2 - 3 | 123 | 139 | ||
| FQ220432.1 | - | 2 - 3 | 133 | 139 | ||
| FQ221142.1 | - | 2 - 3 | 122 | 139 | ||
| FQ221221.1 | - | 2 - 3 | 129 | 139 | ||
| FQ221486.1 | - | 2 - 3 | 125 | 139 | ||
| FQ221683.1 | - | 2 - 3 | 116 | 139 | ||
| FQ221688.1 | - | 2 - 3 | 125 | 139 | ||
| FQ221810.1 | - | 2 - 3 | 125 | 139 | ||
| FQ221946.1 | - | 2 - 3 | 125 | 139 | ||
| FQ221988.1 | - | 2 - 3 | 129 | 139 | ||
| FQ222823.1 | - | 2 - 3 | 125 | 139 | ||
| FQ223282.1 | - | 2 - 3 | 125 | 139 | ||
| FQ224575.1 | - | 2 - 3 | 129 | 139 | ||
| FQ224676.1 | - | 2 - 3 | 125 | 139 | ||
| X78443.1 | - | 2 - 3 | 104 | 139 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl24
Genomic Map