Assay Details
| Gene Symbol: Arf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024152.2 | NP_077066.1 | 1 | 874 | 83 | |
| GenBank mRNA | BC091146.1 | - | 1 | 874 | 7309902 | 83 |
| L12385.1 | - | 1 | 863 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arf6
Genomic Map