Assay Details
| Gene Symbol: Marcksl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030862.2 | NP_110489.1 | 1 | 154 | 77 | |
| GenBank mRNA | AJ301677.1 | - | 1 | 47 | 77 | |
| BC081789.1 | - | 1 | 154 | 7134056 | 77 | |
| Gene Symbol: LOC103689966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759211.1 | XP_008757433.1 | 1 | 188 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ301677.1, BC081789.1, NM_030862.2 transcripts.
Set Membership: |
|

Marcksl1
Genomic Map