Assay Details
| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031670.2 | NP_113858.1 | 3 - 4 | 373 | 80 | |
| GenBank mRNA | AJ251299.1 | - | 3 - 4 | 345 | 80 | |
| BC078790.1 | - | 3 - 4 | 365 | 7109515 | 80 | |
| FQ230345.1 | - | 3 - 4 | 366 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Napsa
Genomic Map