Assay Details
| Gene Symbol: Atp5o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138883.1 | NP_620238.1 | 3 - 4 | 203 | 100 | |
| GenBank mRNA | BC060544.1 | - | 3 - 4 | 211 | 6918775 | 100 |
| D13127.1 | - | 3 - 4 | 203 | 100 | ||
| FQ209940.1 | - | 3 - 4 | 211 | 100 | ||
| FQ210807.1 | - | 3 - 4 | 217 | 100 | ||
| FQ217675.1 | - | 3 - 4 | 208 | 100 | ||
| FQ223905.1 | - | 3 - 4 | 240 | 100 | ||
| FQ226488.1 | - | 3 - 4 | 204 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5o
Genomic Map