Assay Details
| Gene Symbol: Nos1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138922.1 | NP_620277.1 | 7 - 8 | 1145 | 65 | |
| GenBank mRNA | AF037071.1 | - | 7 - 8 | 1145 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nos1ap
Genomic Map