Assay Details
Gene Symbol: Vcpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176857.3 | NP_789827.3 | 2 - 3 | 2906 | 106 | |
GenBank mRNA | AB045378.2 | - | 2 - 3 | 2906 | 106 | |
AF289091.1 | - | 2964 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
