Assay Details
| Gene Symbol: Rab15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198749.2 | NP_942044.1 | 4 - 5 | 697 | 63 | |
| GenBank mRNA | BC094954.1 | - | 4 - 5 | 697 | 7316003 | 63 |
| M83679.1 | - | 4 - 5 | 541 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab15
Genomic Map