Assay Details
| Gene Symbol: Dld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199385.2 | NP_955417.1 | 9 - 10 | 960 | 68 | |
| XM_008764590.1 | XP_008762812.1 | 9 - 10 | 977 | 68 | ||
| GenBank mRNA | BC062069.1 | - | 9 - 10 | 964 | 5599215 | 68 |
| FM046215.1 | - | 1 - 2 | 130 | 68 | ||
| FM093110.1 | - | 4 - 5 | 538 | 68 | ||
| FQ215299.1 | - | 9 - 10 | 967 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dld
Genomic Map