Assay Details
| Gene Symbol: Acsm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144748.1 | NP_653349.1 | 6 - 7 | 998 | 159 | |
| XM_008759677.1 | XP_008757899.1 | 6 - 7 | 983 | 159 | ||
| XM_008759678.1 | XP_008757900.1 | 6 - 7 | 1020 | 159 | ||
| GenBank mRNA | AF062389.1 | - | 6 - 7 | 998 | 159 | |
| BC078721.1 | - | 6 - 7 | 975 | 7110246 | 159 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsm2a
Genomic Map