Assay Details
Gene Symbol: Acad11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108181.1 | NP_001101651.1 | 16 - 17 | 1920 | 61 | |
GenBank mRNA | BC167762.1 | - | 16 - 17 | 1925 | 7443858 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation using acyl-CoA dehydrogenase
long-chain-acyl-CoA dehydrogenase activity
transferase activity, transferring phosphorus-containing groups
very-long-chain-acyl-CoA dehydrogenase activity
flavin adenine dinucleotide binding
medium-chain-acyl-CoA dehydrogenase activity
fatty acid beta-oxidation using acyl-CoA dehydrogenase
long-chain-acyl-CoA dehydrogenase activity
transferase activity, transferring phosphorus-containing groups
very-long-chain-acyl-CoA dehydrogenase activity
flavin adenine dinucleotide binding
medium-chain-acyl-CoA dehydrogenase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P794748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11250 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P794748 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P794748 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P794748 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P441195 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59595 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77278 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10912710 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912707 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912712 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912713 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912715 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912708 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912717 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912704 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912706 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912703 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912711 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912702 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912700 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912714 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912705 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912709 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912701 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912716 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912698 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912699 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10912669 | - | - |
Rat Genome 230 2.0 Array | 1390789_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390789_at | - | - |
Affymetrix HT Rat Focus Array | 1390789_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390789_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800010530.rn.1 | TC0800001444.rn.1 | RTA TC |
Rat Expression Set 230 | 1390789_PM_at | - | - |
Rat Expression Set 230 | 1390789_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912669 | - | - |
Rat Genome U34 Set | rc_AI169321_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22336233 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365021 | - | - |
