Assay Details
Gene Symbol: Pik3r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108777.1 | NP_001102247.1 | 4 - 5 | 1872 | 56 | |
XM_006243800.1 | XP_006243862.1 | 4 - 5 | 1899 | 56 | ||
XM_006243801.2 | XP_006243863.1 | 4 - 5 | 1550 | 56 | ||
GenBank mRNA | BC168984.1 | - | 4 - 5 | 1844 | 7103631 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P520829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66861 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P520829 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P520829 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P520829 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P520829 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73477 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10912772 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912767 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912764 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912759 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912761 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912776 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912774 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912771 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912766 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912765 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912769 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912758 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912768 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912760 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912770 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912762 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912763 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912773 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912775 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10912757 | - | - |
Rat Genome 230 2.0 Array | 1389723_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389723_at | - | - |
Affymetrix HT Rat Focus Array | 1389723_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389723_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800010687.rn.1 | TC0800001463.rn.1 | RTA TC |
Rat Expression Set 230 | 1389723_at | - | - |
Rat Expression Set 230 | 1389723_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912757 | - | - |
Rat Genome U34 Set | rc_AI172265_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22280001 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358726 | - | - |
