Assay Details
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008286.1 | NP_001008287.1 | 3 - 4 | 296 | 70 | |
XM_006234370.2 | XP_006234432.1 | 3 - 4 | 127 | 70 | ||
XM_006234371.2 | XP_006234433.1 | 3 - 4 | 329 | 70 | ||
GenBank mRNA | BC086360.1 | - | 3 - 4 | 296 | 7303884 | 70 |
FQ213350.1 | - | 3 - 4 | 270 | 70 | ||
FQ215122.1 | - | 3 - 4 | 264 | 70 | ||
FQ215507.1 | - | 3 - 4 | 291 | 70 | ||
FQ215582.1 | - | 3 - 4 | 314 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
