Assay Details
| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126288.1 | NP_001119760.1 | 6 - 7 | 813 | 77 | |
| GenBank mRNA | BC161906.1 | - | 6 - 7 | 721 | 8367484 | 77 |
| FQ230808.1 | - | 605 | 77 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Pld4
Genomic Map