Assay Details
| Gene Symbol: Tryx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009173.1 | NP_001009173.1 | 3 - 4 | 461 | 101 | |
| XM_006236368.2 | XP_006236430.1 | 3 - 4 | 1032 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tryx5
Genomic Map