Assay Details
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107858.1 | NP_001101328.1 | 11 - 12 | 1958 | 60 | |
XM_008762883.1 | XP_008761105.1 | 11 - 12 | 1953 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
