Assay Details
Gene Symbol: Slc9b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173973.1 | NP_001167444.1 | 6 - 7 | 1057 | 69 | |
XM_008761518.1 | XP_008759740.1 | 5 - 6 | 573 | 69 | ||
GenBank mRNA | BC158723.1 | - | 1 - 2 | 54 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
