Assay Details
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008771.3 | NP_001008771.2 | 7 - 8 | 787 | 64 | |
GenBank mRNA | AY321348.1 | - | 8 - 9 | 748 | 64 | |
BC097367.1 | - | 7 - 8 | 738 | 7443204 | 64 | |
FQ213175.1 | - | 7 - 8 | 882 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
