Assay Details
| Gene Symbol: Lrif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233147.2 | XP_006233209.1 | 1 - 2 | 167 | 116 | |
| GenBank mRNA | BC158797.1 | - | 1 - 2 | 63 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC158797.1 transcripts.
Set Membership: |
|

Lrif1
Genomic Map