Assay Details
| Gene Symbol: Lrif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127485.1 | NP_001120957.1 | 2 - 3 | 1688 | 140 | |
| XM_008761400.1 | XP_008759622.1 | 2 - 3 | 1522 | 140 | ||
| GenBank mRNA | BC099834.1 | - | 1 - 2 | 105 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P041069 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P041064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121742 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041064 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041069 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041064 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041069 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P041069 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P041064 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P119251 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85131 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13644 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12507 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10818233 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818236 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818231 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818232 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818235 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10818230 | - | - |
| Rat Genome 230 2.0 Array | 1385168_at | - | - |
| Rat Genome 230 2.0 Array | 1385168_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385168_at | - | - |
| Affymetrix HT Rat Focus Array | 1385168_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200013440.rn.1 | TC0200001948.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200013438.rn.1 | TC0200001948.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200013436.rn.1 | TC0200001948.rn.1 | RTA TC |
| Rat Expression Set 230 | 1385168_PM_at | - | - |
| Rat Expression Set 230 | 1385168_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818230 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650018 | - | - |

Lrif1
Genomic Map