Assay Details
| Gene Symbol: Alx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007012.1 | NP_001007013.1 | 2 - 3 | 610 | 68 | |
| GenBank mRNA | AY488087.1 | - | 2 - 3 | 610 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alx3
Genomic Map