Assay Details
Gene Symbol: Clca4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077356.2 | NP_001070824.1 | 7 - 8 | 1217 | 67 | |
GenBank mRNA | AB256513.1 | - | 7 - 8 | 1218 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
