Assay Details
| Gene Symbol: Sdr16c6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109356.1 | NP_001102826.1 | 3 - 4 | 554 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdr16c6| Gene Symbol: Sdr16c6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109356.1 | NP_001102826.1 | 3 - 4 | 554 | 125 | |
Set Membership: |
|