Assay Details
| Gene Symbol: Rgs20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127495.1 | NP_001120967.1 | 3 - 4 | 752 | 69 | |
| GenBank mRNA | BC160842.1 | - | 3 - 4 | 752 | 8361585 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Rgs20
Genomic Map