Assay Details
| Gene Symbol: Sync | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108683.1 | NP_001102153.1 | 3 - 4 | 1451 | 80 | |
| XM_006238943.2 | XP_006239005.1 | 3 - 4 | 1521 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sync
Genomic Map