Assay Details
Gene Symbol: Atpif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012915.2 | NP_037047.2 | 1 - 2 | 131 | 70 | |
GenBank mRNA | BC126065.1 | - | 1 - 2 | 96 | 7442425 | 70 |
D13122.1 | - | 1 - 2 | 131 | 70 | ||
FQ221416.1 | - | 1 - 2 | 116 | 70 | ||
FQ229412.1 | - | 1 - 2 | 93 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
