Assay Details
Gene Symbol: Med18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115037.1 | NP_001108509.1 | 2 - 3 | 293 | 101 | |
GenBank mRNA | BC158672.1 | - | 2 - 3 | 206 | 8371540 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
