Assay Details
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106805.1 | NP_001100275.1 | 13 - 14 | 1872 | 71 | |
| XM_006242630.2 | XP_006242692.2 | 16 - 17 | 2197 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgl3
Genomic Map