Assay Details
| Gene Symbol: Zfp583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228197.2 | XP_006228259.1 | 1 - 2 | 322 | 119 | |
| XM_006228199.2 | XP_006228261.1 | 1 - 2 | 322 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006228197.2, XM_006228199.2 transcripts.
Set Membership: |
|

Zfp583
Genomic Map