Assay Details
Gene Symbol: March10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013973.1 | NP_001013995.1 | 7 - 8 | 2143 | 95 | |
XM_006247579.2 | XP_006247641.1 | 7 - 8 | 2311 | 95 | ||
XM_006247580.2 | XP_006247642.1 | 8 - 9 | 2430 | 95 | ||
GenBank mRNA | AB552845.1 | - | 5 - 6 | 1888 | 95 | |
BC083567.1 | - | 7 - 8 | 2143 | 7112851 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
