Assay Details
| Gene Symbol: Prss22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106984.1 | NP_001100454.1 | 3 - 4 | 358 | 73 | |
| GenBank mRNA | AJ005642.1 | - | 2 - 3 | 342 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss22
Genomic Map