Assay Details
| Gene Symbol: Igfals | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053329.2 | NP_445781.2 | 1 - 2 | 115 | 73 | |
| GenBank mRNA | S46785.1 | - | 1 - 2 | 115 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_053329.2,S46785.1 transcripts.
Set Membership: |
|

Igfals
Genomic Map