Assay Details
| Gene Symbol: Wdr24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191084.1 | NP_001178013.1 | 6 - 7 | 1901 | 61 | |
| XM_008767587.1 | XP_008765809.1 | 6 - 7 | 3207 | 61 | ||
| GenBank mRNA | BC098800.1 | - | 3 - 4 | 397 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr24
Genomic Map